siRNA Details
| Gene Symbol: Ly49i7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009500.1 | NP_001009500.1 | 6 | 703 | ||
| XM_008763320.1 | XP_008761542.1 | 7 | 1800 | |||
| GenBank mRNA | AY659934.1 | - | 6 | 703 | ||
| Gene Symbol: LOC100911272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763440.1 | XP_008761662.1 | 7 | 1559 | ||

Ly49i7
Genomic Map