siRNA Details
| Gene Symbol: Eogt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009502.1 | NP_001009502.1 | 7 | 902 | ||
| XM_008763144.1 | XP_008761366.1 | 4 | 576 | |||
| XM_008763145.1 | XP_008761367.1 | 5 | 488 | |||
| XM_008763146.1 | XP_008761368.1 | 3 | 368 | |||
| XM_008763147.1 | XP_008761369.1 | 4 | 633 | |||
| GenBank mRNA | AJ868236.1 | - | 7 | 902 | ||

Eogt
Genomic Map