siRNA Details
| Gene Symbol: Eogt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009502.1 | NP_001009502.1 | 11 | 1310 | ||
| XM_008763144.1 | XP_008761366.1 | 8 | 984 | |||
| XM_008763145.1 | XP_008761367.1 | 9 | 896 | |||
| XM_008763146.1 | XP_008761368.1 | 7 | 776 | |||
| XM_008763147.1 | XP_008761369.1 | 8 | 1041 | |||
| GenBank mRNA | AJ868236.1 | - | 11 | 1310 | ||

Eogt
Genomic Map