siRNA Details
| Gene Symbol: Vom1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009510.1 | NP_001009510.1 | 1 | 712 | ||
| GenBank mRNA | AY510339.1 | - | 1 | 712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r21| Gene Symbol: Vom1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009510.1 | NP_001009510.1 | 1 | 712 | ||
| GenBank mRNA | AY510339.1 | - | 1 | 712 | ||