siRNA Details
| Gene Symbol: B4galt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009539.1 | NP_001009539.1 | 3 | 410 | ||
| XM_006250253.2 | XP_006250315.1 | 3 | 441 | |||
| XM_006250254.2 | XP_006250316.1 | 3 | 561 | |||
| GenBank mRNA | BC061812.1 | - | 3 | 410 | 5598658 | |

B4galt3
Genomic Map