siRNA Details
| Gene Symbol: CDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001802.1 | NP_001793.1 | 5 | 2380 | ||
| GenBank mRNA | BC017503.1 | - | 5 | 2364 | 4873337 | |
| BM714067.1 | - | 1 | 239 | |||
| M63256.1 | - | 5 | 2238 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDR2| Gene Symbol: CDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001802.1 | NP_001793.1 | 5 | 2380 | ||
| GenBank mRNA | BC017503.1 | - | 5 | 2364 | 4873337 | |
| BM714067.1 | - | 1 | 239 | |||
| M63256.1 | - | 5 | 2238 | |||