siRNA Details
| Gene Symbol: CDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001802.1 | NP_001793.1 | 3 | 547 | ||
| GenBank mRNA | AK292273.1 | - | 3 | 294 | ||
| BC017503.1 | - | 3 | 529 | 4873337 | ||
| BG750955.1 | - | 3 | 530 | |||
| D12981.1 | - | 3 | 223 | |||
| M63256.1 | - | 3 | 407 | |||

CDR2
Genomic Map