siRNA Details
| Gene Symbol: NOP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291978.1 | NP_001278907.1 | 1, 2 | 390 | ||
| NM_001291979.1 | NP_001278908.1 | 1, 2 | 390 | |||
| NM_003703.2 | NP_003694.1 | 1, 2 | 390 | |||
| GenBank mRNA | AF040965.1 | - | 1, 2 | 252 | ||
| AK292939.1 | - | 1, 2 | 390 | |||
| BC026035.1 | - | 1, 2 | 217 | 4878317 | ||
| BC053635.1 | - | 1, 2 | 269 | 5114280 | ||

NOP14
Genomic Map