siRNA Details
Gene Symbol: Eln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012722.1 | NP_036854.1 | 37 | 3371 | ||
GenBank mRNA | BC085910.1 | - | 37 | 3371 | 7097661 | |
J04035.1 | - | 6 | 993 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012722.1 | NP_036854.1 | 37 | 3371 | ||
GenBank mRNA | BC085910.1 | - | 37 | 3371 | 7097661 | |
J04035.1 | - | 6 | 993 |