siRNA Details
| Gene Symbol: WSCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304447.1 | NP_001291376.1 | 10 | 3691 | ||
| NM_014653.3 | NP_055468.2 | 9 | 3860 | |||
| XM_017020243.1 | XP_016875732.1 | 10 | 4145 | |||
| XM_017020244.1 | XP_016875733.1 | 11 | 4306 | |||
| XM_017020245.1 | XP_016875734.1 | 11 | 3803 | |||
| XM_017020246.1 | XP_016875735.1 | 9 | 3534 | |||
| XM_017020247.1 | XP_016875736.1 | 10 | 3677 | |||
| XM_017020248.1 | XP_016875737.1 | 10 | 4149 | |||
| XM_017020249.1 | XP_016875738.1 | 9 | 4006 | |||
| XM_017020250.1 | XP_016875739.1 | 7 | 3560 | |||
| XM_017020251.1 | XP_016875740.1 | 8 | 3371 | |||
| XM_017020252.1 | XP_016875741.1 | 8 | 3849 | |||
| GenBank mRNA | AB018332.2 | - | 9 | 3597 | ||
| AK098069.1 | - | 1 | 1392 | |||
| BC037969.1 | - | 5 | 2146 | |||
| BC110330.1 | - | 9 | 3643 | 5735001 | ||
| BC136687.1 | - | 8 | 3389 | 9020677 | ||
| BC136688.1 | - | 8 | 3389 | 9020678 | ||
| BX647283.1 | - | 10 | 3642 | |||

WSCD2
Genomic Map