siRNA Details
| Gene Symbol: KIAA0100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014680.3 | NP_055495.2 | 4 | 479 | ||
| XM_005258073.2 | XP_005258130.1 | 4 | 474 | |||
| GenBank mRNA | AB621823.1 | - | 4 | 476 | ||
| AK130007.1 | - | 4 | 434 | |||
| AK307216.1 | - | 3 | 418 | |||
| AY943906.1 | - | 1 | 324 | |||
| BC048096.1 | - | 3 | 729 | |||
| BC050440.1 | - | 4 | 475 | |||
| BC120873.1 | - | 4 | 338 | 40075102 | ||
| CR933634.1 | - | 4 | 431 | |||
| D43947.3 | - | 4 | 451 | |||

KIAA0100
Genomic Map