siRNA Details
| Gene Symbol: KIAA0100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014680.3 | NP_055495.2 | 17 | 3202 | ||
| XM_005258073.2 | XP_005258130.1 | 17 | 3197 | |||
| XM_017025423.1 | XP_016880912.1 | 16 | 3051 | |||
| GenBank mRNA | BC120873.1 | - | 17 | 3061 | 40075102 | |
| CR933634.1 | - | 18 | 3064 | |||
| D43947.3 | - | 17 | 3174 | |||

KIAA0100
Genomic Map