siRNA Details
| Gene Symbol: RUSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135999.1 | NP_001129471.1 | 2 | 2008 | ||
| NM_014806.3 | NP_055621.1 | 2 | 1752 | |||
| NR_052015.2 | 2 | 1752 | ||||
| GenBank mRNA | AB002373.2 | - | 2 | 1594 | ||
| AK299805.1 | - | 2 | 1698 | |||
| BC064843.1 | - | 1 | 796 | |||
| BC132766.1 | - | 1 | 1537 | 40146788 | ||
| BC132770.1 | - | 1 | 1537 | 40146792 | ||
| BC146654.1 | - | 2 | 1726 | 40147826 | ||
| BC150211.1 | - | 2 | 1664 | |||
| BC150262.1 | - | 2 | 1605 | 9007205 | ||

RUSC2
Genomic Map