siRNA Details
| Gene Symbol: SLC8A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015063.2 | NP_055878.1 | 10 | 3869 | ||
| XM_005259172.1 | XP_005259229.1 | 9 | 3660 | |||
| XM_017027159.1 | XP_016882648.1 | 7 | 2017 | |||
| GenBank mRNA | AB029010.1 | - | 10 | 3684 | ||
| AL512764.1 | - | 9 | 3058 | |||

SLC8A2
Genomic Map