siRNA Details
| Gene Symbol: TMEM63B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318792.1 | NP_001305721.1 | 12 | 1109 | ||
| NM_018426.2 | NP_060896.1 | 12 | 1075 | |||
| XM_005249211.3 | XP_005249268.1 | 14 | 1455 | |||
| XM_005249213.4 | XP_005249270.1 | 13 | 1382 | |||
| XM_005249214.4 | XP_005249271.1 | 11 | 969 | |||
| XM_005249215.4 | XP_005249272.1 | 12 | 1274 | |||
| XM_005249217.1 | XP_005249274.1 | 12 | 1147 | |||
| XM_006715135.1 | XP_006715198.1 | 12 | 1131 | |||
| XM_017010999.1 | XP_016866488.1 | 14 | 1332 | |||
| XM_017011000.1 | XP_016866489.1 | 13 | 1313 | |||
| XM_017011001.1 | XP_016866490.1 | 11 | 1013 | |||
| XM_017011002.1 | XP_016866491.1 | 11 | 1116 | |||
| GenBank mRNA | AK093629.1 | - | 12 | 1131 | ||
| AL834238.1 | - | 5 | 373 | |||
| BC022095.1 | - | 9 | 712 | |||
| BC136769.1 | - | 11 | 951 | 9020759 | ||

TMEM63B
Genomic Map