siRNA Details
| Gene Symbol: HEATR5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019024.2 | NP_061897.1 | 35 | 5951 | ||
| XM_006712034.2 | XP_006712097.1 | 35 | 5887 | |||
| XM_006712035.3 | XP_006712098.1 | 35 | 5954 | |||
| XM_011532934.2 | XP_011531236.1 | 35 | 6418 | |||
| XM_011532935.2 | XP_011531237.1 | 24 | 4090 | |||
| XM_017004378.1 | XP_016859867.1 | 34 | 5623 | |||
| XM_017004379.1 | XP_016859868.1 | 36 | 6328 | |||
| XM_017004380.1 | XP_016859869.1 | 36 | 6101 | |||
| GenBank mRNA | AB037835.1 | - | 26 | 4398 | ||
| AK026928.1 | - | 12 | 2211 | |||
| AL832880.1 | - | 8 | 1353 | |||
| BC150605.1 | - | 35 | 5936 | 9056975 | ||
| BX538008.1 | - | 29 | 4874 | |||
| BX648805.1 | - | 21 | 4111 | |||

HEATR5B
Genomic Map