siRNA Details
| Gene Symbol: KIDINS220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020738.2 | NP_065789.1 | 5 | 533 | ||
| GenBank mRNA | AB033076.2 | - | 5 | 508 | ||
| AB621829.1 | - | 5 | 521 | |||
| AK057125.1 | - | 5 | 521 | |||
| AK296459.1 | - | 5 | 520 | |||
| AL133620.1 | - | 5 | 508 | |||
| BC130610.1 | - | 6 | 628 | 40146641 | ||
| DA507846.1 | - | 5 | 533 | |||

KIDINS220
Genomic Map