siRNA Details
| Gene Symbol: KIDINS220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020738.2 | NP_065789.1 | 12 | 1400 | ||
| GenBank mRNA | AB033076.2 | - | 12 | 1375 | ||
| AK057125.1 | - | 12 | 1388 | |||
| AK294832.1 | - | 8 | 704 | |||
| AK296459.1 | - | 12 | 1390 | |||
| AL133620.1 | - | 12 | 1378 | |||
| BC130610.1 | - | 13 | 1495 | 40146641 | ||

KIDINS220
Genomic Map