siRNA Details
| Gene Symbol: KLHL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020782.1 | NP_065833.1 | 3 | 3005 | ||
| XM_017019698.1 | XP_016875187.1 | 3 | 3770 | |||
| GenBank mRNA | AB037761.1 | - | 3 | 2736 | ||
| BC137116.1 | - | 3 | 2972 | 9021113 | ||
| BC137117.1 | - | 3 | 2971 | 9021114 | ||
| T83155.1 | - | 1 | 164 | |||

KLHL42
Genomic Map