siRNA Details
| Gene Symbol: CEP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020802.3 | NP_065853.3 | 11 | 3869 | ||
| XM_005271627.4 | XP_005271684.2 | 11 | 3568 | |||
| GenBank mRNA | AB037798.1 | - | 8 | 3212 | ||
| AK095004.1 | - | 10 | 3499 | |||
| AK296350.1 | - | 8 | 3610 | |||

CEP126
Genomic Map