siRNA Details
| Gene Symbol: KIAA1586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286274.1 | NP_001273203.1 | 3 | 1826 | ||
| NM_001286275.1 | NP_001273204.1 | 4 | 1885 | |||
| NM_001286276.1 | NP_001273205.1 | 4 | 1837 | |||
| NM_020931.3 | NP_065982.1 | 4 | 1907 | |||
| GenBank mRNA | AB046806.1 | - | 1 | 3019 | ||
| AK290988.1 | - | 4 | 1701 | |||
| AK295519.1 | - | 3 | 1632 | |||
| BC041125.1 | - | 4 | 1853 | 5785266 | ||

KIAA1586
Genomic Map