siRNA Details
| Gene Symbol: CWC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020943.2 | NP_065994.1 | 7 | 957 | ||
| XM_005246726.2 | XP_005246783.1 | 7 | 1060 | |||
| GenBank mRNA | AB046824.1 | - | 6 | 776 | ||
| AK025635.1 | - | 7 | 814 | |||
| BC016651.1 | - | 6 | 781 | |||
| BC031216.1 | - | 7 | 957 | |||
| BC053573.1 | - | 4 | 479 | |||
| BC057826.1 | - | 7 | 815 | |||
| BC093952.1 | - | 6 | 679 | 7939797 | ||
| BC093954.1 | - | 6 | 679 | 7939799 | ||
| BC143433.1 | - | 6 | 678 | |||

CWC22
Genomic Map