siRNA Details
| Gene Symbol: CWC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020943.2 | NP_065994.1 | 20 | 3200 | ||
| XM_005246726.2 | XP_005246783.1 | 20 | 3303 | |||
| GenBank mRNA | AB046824.1 | - | 19 | 3019 | ||
| AK026978.1 | - | 13 | 2157 | |||
| AK123647.1 | - | 7 | 2157 | |||
| BC041662.1 | - | 13 | 2151 | |||
| BC053573.1 | - | 17 | 2722 | |||

CWC22
Genomic Map