siRNA Details
| Gene Symbol: Gnas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024823.4 | NP_001019994.4 | 1 | 2188 | ||
| NM_021845.5 | NP_068617.6 | 1 | 2188 | |||
| GenBank mRNA | X84047.1 | - | 1 | 1336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnas| Gene Symbol: Gnas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024823.4 | NP_001019994.4 | 1 | 2188 | ||
| NM_021845.5 | NP_068617.6 | 1 | 2188 | |||
| GenBank mRNA | X84047.1 | - | 1 | 1336 | ||