siRNA Details
| Gene Symbol: ZYG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024646.2 | NP_078922.1 | 14 | 5763 | ||
| XM_006710898.3 | XP_006710961.1 | 14 | 5650 | |||
| GenBank mRNA | AB051517.1 | - | 14 | 5713 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZYG11B| Gene Symbol: ZYG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024646.2 | NP_078922.1 | 14 | 5763 | ||
| XM_006710898.3 | XP_006710961.1 | 14 | 5650 | |||
| GenBank mRNA | AB051517.1 | - | 14 | 5713 | ||