siRNA Details
| Gene Symbol: ERMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024896.2 | NP_079172.2 | 15 | 3914 | ||
| XM_005251587.1 | XP_005251644.1 | 13 | 3519 | |||
| XM_017015139.1 | XP_016870628.1 | 16 | 3707 | |||
| GenBank mRNA | AB058718.2 | - | 15 | 3791 | ||
| AK026962.1 | - | 7 | 2220 | |||
| AL834191.1 | - | 14 | 3546 | |||

ERMP1
Genomic Map