siRNA Details
| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026601.3 | NP_080877.3 | 4 | 567 | ||
| GenBank mRNA | AK004328.1 | - | 3 | 155 | ||
| AK020099.1 | - | 4 | 441 | |||
| BC024619.1 | - | 4 | 555 | 4923664 | ||
| BC100401.1 | - | 4 | 531 | |||
| CB952524.1 | - | 2 | 72 | |||
| CK620463.1 | - | 4 | 455 | |||

Hyi
Genomic Map