siRNA Details
| Gene Symbol: Ccnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028399.1 | NP_082675.1 | 3 | 417 | ||
| GenBank mRNA | AK013634.1 | - | 3 | 364 | ||
| AK036772.1 | - | 365 | ||||
| AK049863.1 | - | 3 | 362 | |||
| AK152399.1 | - | 3 | 365 | |||
| AK154439.1 | - | 3 | 375 | |||
| AK156231.1 | - | 386 | ||||
| AK156621.1 | - | 3 | 373 | |||
| BC054122.1 | - | 3 | 417 | 6509024 | ||
| JX965957.1 | - | 3 | 353 | |||
| JX965958.1 | - | 3 | 353 | |||

Ccnt2
Genomic Map