siRNA Details
| Gene Symbol: Ccnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028399.1 | NP_082675.1 | 9 | 2027 | ||
| GenBank mRNA | AK047125.1 | - | 5 | 2927 | ||
| AK152399.1 | - | 9 | 1975 | |||
| AK154439.1 | - | 9 | 1985 | |||
| BC054122.1 | - | 9 | 2027 | 6509024 | ||
| JX965958.1 | - | 9 | 1963 | |||

Ccnt2
Genomic Map