siRNA Details
| Gene Symbol: SPHKAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142644.1 | NP_001136116.1 | 12 | 6854 | ||
| NM_030623.3 | NP_085126.2 | 11 | 6767 | |||
| XM_005246870.4 | XP_005246927.1 | 13 | 7326 | |||
| XM_006712777.3 | XP_006712840.1 | 13 | 7287 | |||
| XM_006712778.3 | XP_006712841.1 | 12 | 7199 | |||
| XM_011511925.2 | XP_011510227.1 | 14 | 7405 | |||
| GenBank mRNA | AB051465.1 | - | 5 | 5615 | ||
| AK095042.1 | - | 6 | 3016 | |||
| AK125481.1 | - | 1 | 3397 | |||
| CR749297.1 | - | 6 | 5960 | |||
| CR749494.1 | - | 11 | 6749 | |||

SPHKAP
Genomic Map