siRNA Details
| Gene Symbol: Aoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031582.2 | NP_113770.2 | 1 | 762 | ||
| GenBank mRNA | AB195675.1 | - | 1 | 767 | ||
| BC100613.1 | - | 1 | 762 | 5598211 | ||
| U72632.1 | - | 1 | 767 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aoc3| Gene Symbol: Aoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031582.2 | NP_113770.2 | 1 | 762 | ||
| GenBank mRNA | AB195675.1 | - | 1 | 767 | ||
| BC100613.1 | - | 1 | 762 | 5598211 | ||
| U72632.1 | - | 1 | 767 | |||