siRNA Details
| Gene Symbol: Col3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032085.1 | NP_114474.1 | 40, 41 | 3040 | ||
| GenBank mRNA | AJ005395.1 | - | 2, 3 | 240 | ||
| BC087039.1 | - | 40, 41 | 3040 | 7097081 | ||
| X70369.1 | - | 5, 6 | 456 | |||

Col3a1
Genomic Map