siRNA Details
| Gene Symbol: TMEM132B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286219.1 | NP_001273148.1 | 5 | 1199 | ||
| NM_052907.3 | NP_443139.2 | 9 | 2452 | |||
| XM_011537852.2 | XP_011536154.1 | 9 | 2509 | |||
| XM_011537854.2 | XP_011536156.1 | 5 | 7182 | |||
| XM_017018767.1 | XP_016874256.1 | 10 | 2618 | |||
| GenBank mRNA | AB067493.1 | - | 4 | 989 | ||
| AK093097.1 | - | 5 | 1155 | |||
| BC113366.1 | - | 5 | 1076 | 8322418 | ||
| BC113368.1 | - | 5 | 1076 | 8322420 | ||
| BC131621.1 | - | 5 | 1116 | 40117411 | ||

TMEM132B
Genomic Map