siRNA Details
| Gene Symbol: Spp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053577.1 | NP_446029.1 | 337 | |||
| GenBank mRNA | BC088193.1 | - | 337 | 7372900 | ||
| FQ210158.1 | - | 253 | ||||
| FQ210198.1 | - | 274 | ||||
| FQ210350.1 | - | 301 | ||||
| U19485.1 | - | 118 | ||||

Spp2
Genomic Map