siRNA Details
| Gene Symbol: Idh3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053581.1 | NP_446033.1 | 12 | 1204 | ||
| GenBank mRNA | AB047540.1 | - | 5 | 473 | ||
| BC079113.1 | - | 12 | 1204 | 7126029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Idh3B| Gene Symbol: Idh3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053581.1 | NP_446033.1 | 12 | 1204 | ||
| GenBank mRNA | AB047540.1 | - | 5 | 473 | ||
| BC079113.1 | - | 12 | 1204 | 7126029 | ||