siRNA Details
| Gene Symbol: Ppp2r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057140.1 | NP_476481.1 | 15 | 2002 | ||
| GenBank mRNA | BC083859.1 | - | 15 | 2002 | 7126254 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r1a| Gene Symbol: Ppp2r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057140.1 | NP_476481.1 | 15 | 2002 | ||
| GenBank mRNA | BC083859.1 | - | 15 | 2002 | 7126254 | |