siRNA Details
| Gene Symbol: FNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008738.2 | NP_001008738.2 | 15 | 3265 | ||
| NM_133372.2 | NP_588613.2 | 16 | 3349 | |||
| GenBank mRNA | AB075841.1 | - | 10 | 2538 | ||
| AK126675.1 | - | 3 | 709 | |||
| AK292473.1 | - | 16 | 3303 | |||
| AL832008.1 | - | 3 | 1361 | |||
| AL834127.2 | - | 3 | 1427 | |||
| CR749426.1 | - | 11 | 2565 | |||
| CR936723.1 | - | 16 | 3079 | |||
| DQ145719.1 | - | 16 | 3254 | |||

FNIP1
Genomic Map