siRNA Details
| Gene Symbol: KIAA1958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287036.1 | NP_001273965.1 | 5 | 2109 | ||
| NM_001287038.1 | NP_001273967.1 | 4 | 2015 | |||
| NM_133465.3 | NP_597722.1 | 4 | 2025 | |||
| GenBank mRNA | AB075838.1 | - | 3 | 1525 | ||
| AK056181.1 | - | 3 | 1237 | |||
| AK096613.1 | - | 4 | 2026 | |||
| BC105048.1 | - | 4 | 1915 | 8144051 | ||
| BC113515.1 | - | 4 | 1915 | 8322567 | ||
| BC143424.1 | - | 5 | 1999 | 9051932 | ||

KIAA1958
Genomic Map