siRNA Details
| Gene Symbol: SOGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012279.2 | NP_001012279.1 | 12 | 7697 | ||
| Gene Symbol: KIAA0408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014702.4 | NP_055517.3 | 6 | 4003 | ||
| GenBank mRNA | AB007868.1 | - | 4 | 4396 | ||
| AK127114.1 | - | 2 | 1970 | |||

SOGA3
Genomic Map