siRNA Details
| Gene Symbol: Pkdcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134117.2 | NP_598878.2 | 6, 7 | 1584 | ||
| GenBank mRNA | AB381893.1 | - | 6, 7 | 1576 | ||
| AB436780.1 | - | 6, 7 | 1565 | |||
| AK133913.1 | - | 2, 3 | 261 | |||
| BC022157.1 | - | 5, 6 | 731 | |||
| BC086639.1 | - | 7, 8 | 1422 | 6828629 | ||

Pkdcc
Genomic Map