siRNA Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134364.1 | NP_599191.1 | 10 | 1702 | ||
| GenBank mRNA | BC099743.1 | - | 10 | 1685 | 7937989 | |
| FQ214851.1 | - | 7 | 1194 | |||
| FQ216116.1 | - | 8 | 1284 | |||
| FQ230301.1 | - | 8 | 1321 | |||
| FQ231816.1 | - | 6 | 1081 | |||
| M19044.1 | - | 9 | 1518 | |||
| M57634.1 | - | 7 | 1166 | |||

Atp5b
Genomic Map