siRNA Details
| Gene Symbol: GLCCI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138426.3 | NP_612435.1 | 8 | 2408 | ||
| GenBank mRNA | AK055741.1 | - | 1 | 1809 | ||
| AK126731.1 | - | 2 | 1559 | |||
| AL832860.1 | - | 2 | 520 | |||
| AL833694.1 | - | 4 | 903 | |||
| BC011254.1 | - | 2 | 616 | |||
| BC050291.1 | - | 8 | 2358 | 5272003 | ||

GLCCI1
Genomic Map