siRNA Details
| Gene Symbol: ZNF700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271848.1 | NP_001258777.1 | 4 | 687 | ||
| NM_144566.2 | NP_653167.1 | 4 | 678 | |||
| GenBank mRNA | AK126871.1 | - | 3 | 899 | ||
| AK308305.1 | - | 4 | 664 | |||
| AL136732.1 | - | 4 | 661 | |||
| BC136777.1 | - | 4 | 605 | 9020767 | ||
| BC144406.1 | - | 5 | 726 | |||

ZNF700
Genomic Map