siRNA Details
| Gene Symbol: AFAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146337.2 | NP_001139809.1 | 18 | 3045 | ||
| NM_001323062.1 | NP_001309991.1 | 18 | 3075 | |||
| NM_152406.3 | NP_689619.1 | 19 | 3174 | |||
| XM_011537558.1 | XP_011535860.1 | 15 | 2724 | |||
| XM_017009036.1 | XP_016864525.1 | 19 | 3151 | |||
| GenBank mRNA | AK074185.1 | - | 19 | 3117 | ||
| AK094067.1 | - | 10 | 2204 | |||

AFAP1L1
Genomic Map