siRNA Details
Gene Symbol: Vom2r31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173319.1 | NP_775441.1 | 6 | 3747 | ||
GenBank mRNA | AF016184.1 | - | 6 | 3747 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
