siRNA Details
| Gene Symbol: Tbc1d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194334.2 | NP_919315.2 | 13 | 4745 | ||
| GenBank mRNA | AK129273.1 | - | 13 | 4730 | ||
| BC030847.1 | - | 3 | 2154 | |||
| BC056467.1 | - | 6 | 3056 | |||
| BC108420.1 | - | 8 | 3350 | |||
| CB056522.1 | - | 1 | 127 | |||

Tbc1d2b
Genomic Map