siRNA Details
| Gene Symbol: Ly6g5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001934.1 | NP_001001934.1 | 3 | 263 | ||
| XM_008772747.1 | XP_008770969.1 | 4 | 950 | |||
| GenBank mRNA | HE864428.1 | - | 2 | 174 | ||
| HE864429.1 | - | 3 | 303 | |||
| Gene Symbol: Csnk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | HE864431.1 | - | 546 | |||
| HE864432.1 | - | 1695 | ||||
| HE864433.1 | - | 1920 | ||||
| HE864434.1 | - | 2176 | ||||

Ly6g5b
Genomic Map