siRNA Details
| Gene Symbol: Utp14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001981.3 | NP_001001981.2 | 3 | 3579 | ||
| NM_001136226.1 | NP_001129698.1 | 4 | 3586 | |||
| GenBank mRNA | AK029972.1 | - | 4 | 3589 | ||
| AK046254.1 | - | 1 | 1495 | |||
| AY316161.1 | - | 3 | 3365 | |||
| AY316162.1 | - | 1 | 1613 | |||
| BC039778.1 | - | 1 | 2325 | |||

Utp14b
Genomic Map