siRNA Details
| Gene Symbol: Pi4ka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001983.2 | NP_001001983.2 | 37, 38 | 4320 | ||
| GenBank mRNA | AK136942.1 | - | 1, 2 | 107 | ||
| BC075629.1 | - | 37, 38 | 4320 | 6856662 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pi4ka| Gene Symbol: Pi4ka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001983.2 | NP_001001983.2 | 37, 38 | 4320 | ||
| GenBank mRNA | AK136942.1 | - | 1, 2 | 107 | ||
| BC075629.1 | - | 37, 38 | 4320 | 6856662 | ||