siRNA Details
| Gene Symbol: Kdm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001984.2 | NP_001001984.2 | 12 | 2212 | ||
| GenBank mRNA | AK155866.1 | - | 12 | 2217 | ||
| AK173084.1 | - | 1 | 30 | |||
| BC037117.1 | - | 5 | 761 | |||
| BC076576.1 | - | 12 | 1778 | 6840591 | ||

Kdm2a
Genomic Map